4-Methylmorpholine for synthesis
Supplier: Merck
Certificates
About this item
Synonyms: NMM, N-Methylmorpholine
CAS Number:109-02-4
:
Formula:C₅H₁₁NO
MDL Number:MFCD00006175
Boiling Pt:115
Melting Pt:–66 °C
Density:0.922 g/cm³ (20 °C)
Molecular Weight:101.15 g/mol
Flash Pt:12
Storage Temperature:Ambient
UN:2535
ADR:3 (8) II
:M-Clarity™ quality level = MQ200
EINECS:203-640-0
:GHS02
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together