Order Entry
Ireland
ContactUsLinkComponent
 
2-Methoxycinnamic acid
Catalog # MOLEM50845282
Supplier:  Molekula
CAS Number:  
2-Methoxycinnamic acid
Catalog # MOLEM50845282
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3-(2-Methoxyphenyl)acrylic acid, 2-Methoxycinnamic Acid
 :
 :178.18 g/mol
EINECS:228-047-4
 :GHS07
CAS Number:6099-03-2
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00064238