Order Entry
Ireland
ContactUsLinkComponent
Sodium oleate
Sodium oleate
Catalog # ROTH8856.1
Supplier:  Roth Carl
CAS Number:  
Sodium oleate
Catalog # ROTH8856.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Melting Pt:232…235 °C
Density:1,02 g/cm³ (20 °C)
 :304.45 g/mol
Storage Temperature:Freezer
EINECS:205-591-0
CAS Number:143-19-1
Formula:C₁₈H₃₃NaO₂
MDL Number:MFCD00004438