HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid)
Catalog # MOLE47376574-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Merck Index:13,04672
Melting Pt:210…215 °C
Density:1,448 g/cm³ (20 °C)
:238.31 g/mol
Storage Temperature:Ambient
EINECS:230-907-9
CAS Number:7365-45-9
Formula:C₈H₁₈N₂O₄S
MDL Number:MFCD00006158