(4-Bromophenyl)acetic acid
Glass bottle for solids, 100 g
Catalog # MOLEM79536365
Marketsource
Supplier: Molekula
(4-Bromophenyl)acetic acid
Catalog # MOLEM79536365
Supplier: Molekula
Marketsource
CAS Number: 1878-68-8
Jump to:
Product Details & Documents
Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid
- CAS Number:1878-68-8
- Formula:(C₆H₄Br)CH₂COOH
- MDL Number:MFCD00004342
- Molecular Weight:215.05 g/mol
- Boiling Pt:326 °C (1013 hPa)
- Melting Pt:94…98 °C
- EINECS:217-523-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



