Order Entry
Ireland
ContactUsLinkComponent
tert-Butanol ≥99%, GPR RECTAPUR®
Flammable
Irritant
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Density (26/4):
    0.770 - 0.780
  • Solidification point:
    23.5 - 26.5 °C
  • Evaporation residue:
    Max. 100 ppm
  • Water:
    Max. 0.5 %

Specification Test Results

Frequently Bought Together