Order Entry
Ireland
ContactUsLinkComponent
 
 
 
 
Isobutanol ≥97%, TECHNICAL
  20835.294
   Safer Human and Environmental Health
 :  
undefined
 
 
 
Isobutanol ≥97%, TECHNICAL
  20835.294
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Assay:
    Min. 97 %
  • Refractive index (20°C):
    1.395 - 1.396

 

 

 :
 :74.12 g/mol
 :13,05148
 :107,9 °C (1013 hPa)
 :–108 °C
 :0,802 g/cm³ (20 °C)
 :28 °C
 :201-148-0
 :1212
 :3,III
 :GHS02
 :GHS05
 :GHS07
 :Ambient
 :78-83-1
 :(CH₃)₂CHCH₂OH
 :MFCD00004740