
1-Octanesulphonic acid sodium salt
Catalog # MOLE18122348-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:216.28 g/mol
Melting Pt:> 300 °C
EINECS:226-195-4
:GHS07
Storage Temperature:Ambient
CAS Number:5324-84-5
Formula:C₈H₁₇NaO₃S
MDL Number:MFCD00007544