1-Hexanesulphonic acid sodium salt
: Molekula
:2832-45-3
:MFCD00007542
:C₆H₁₃O₃SNa
:300 °C
:188.22 g/mol
:Ambient
:220-601-3
Frequently Bought Together
Cryoboxes, Nalgene®
: Thermo Fisher Scientific
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Sampling Spoons
: VWR Collection
Cell culture plates, Nunc™ Multidish
: Thermo Fisher Scientific
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Storage plates, 96-well, MASTERBLOCK®
: Greiner Bio-One