1-Butanesulphonic acid sodium salt
Supplier: Molekula
Certificates
About this item
CAS Number:2386-54-1
:
Formula:C₄H₉NaO₃S
MDL Number:MFCD00007540
:160.17 g/mol
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:219-201-1
Frequently Bought Together