Lithium dodecyl sulphate
Catalog # MOLE14603001-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:272.34 g/mol
EINECS:218-058-2
:GHS07
Storage Temperature:Ambient
CAS Number:2044-56-6
Formula:C₁₂H₂₅LiO₄S
MDL Number:MFCD00007467