Order Entry
Ireland
ContactUsLinkComponent
 
 
Dithiothreitol (DTT, Cleland's reagent) 99+% for molecular biology DNase-, RNase-, protease-free
 
undefined
 
Dithiothreitol (DTT, Cleland's reagent) 99+% for molecular biology DNase-, RNase-, protease-free
 

 

  • Appearance:
    White to off-white Solid
  • Infrared spectrum:
    Conforms
  • Titration Iodometric:
    99.0 to 101.0 %
  • UV:
    at 260 nm A: ≤0.5
  • Dnase,Rnase,Protease activity:
    none detected
  • Heavy metals (as Pb):
    ≤5 ppm
  • Oxidized DTT(UV):
    ≤0.5 %

 

 

Frequently Bought Together