

1,4-Diaminobutane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-60-1
:
Formula:C₄H₁₂N₂
MDL Number:MFCD00008235
:88.15 g/mol
Merck Index:14,7947
Boiling Pt:158…160 °C
Melting Pt:25…28 °C
Density:0,877
Flash Pt:63 °C (145 °F)
Storage Temperature:Ambient
EINECS:203-782-3
UN:2928
ADR:6.1,II
:GHS06
:GHS05
Related Information
Frequently Bought Together