Order Entry
Ireland
ContactUsLinkComponent
Taurine for synthesis, Sigma-Aldrich®
Taurine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Taurine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    65 g/l (12 °C)
  • Melting Point:
    300 °C (decomposition)
  • Molar Mass:
    125.15 g/mol

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together