
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Supplier: Roth Carl
Certificates
About this item
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1,328 g/cm³ (20 °C)
EINECS:201-064-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together