Order Entry
Ireland
ContactUsLinkComponent
3-Aminopropan-1-ol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Corrosive
Irritant
3-Aminopropan-1-ol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
3-Aminopropan-1-ol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Assay (GC, area%):
    ≥ 99.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    0.986 - 0.988
  • Water (K. F.):
    ≤ 0.30 %
  • Identity (IR):
    passes test

Specification Test Results

Related Information

Frequently Bought Together