

3-Aminopropan-1-ol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:156-87-6
:
Formula:C₃H₉NO
MDL Number:MFCD00008223
:75.11 g/mol
Boiling Pt:187…188 °C (760 mmHg)
Melting Pt:10…12 °C
Density:0,982 g/cm³
Flash Pt:101 °C
Storage Temperature:Ambient
EINECS:205-864-4
UN:2735
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4595 to 1.4615 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together