

Ethanolamine, EMPROVE® EXPERT BP, NF for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:141-43-5
:
Formula:NH₂CH₂CH₂OH
MDL Number:MFCD00008183
Merck Index:13,03762
Boiling Pt:171 °C (1013 hPa)
Melting Pt:10,5 °C
Density:1,0164 g/cm³ (20 °C)
Flash Pt:93 °C
UN:2491
ADR:8,
:61.08 g/mol
EINECS:205-483-3
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together