Order Entry
Ireland
ContactUsLinkComponent
 
Triethanolamine (trolamine) ≥95%, TECHNICAL
  28743.290
 :  
undefined
Triethanolamine (trolamine) ≥95%, TECHNICAL
  28743.290
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay:
    Min. 95 %

 

 

 :
 :149.19 g/mol
 :13,09739
 :335,4 °C (1013 hPa)
 :21,6 °C
 :1,13 g/cm³ (20 °C)
 :179 °C (closed cup)
 :Ambient
 :203-049-8
 :102-71-6
 :C₆H₁₅NO₃
 :MFCD00002855