


N,N-Dimethylformamide dimethyl acetal ca. 97%
:4637-24-5
:
:C₅H₁₃NO₂
:MFCD00008482
:119.16 g/mol
:102 °C (1013 hPa)
:0,89 g/cm³
:7 °C
:225-063-3
:1993
:GHS07
:GHS02
:GHS05
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3970 (20°C, 589 nm)
- GC:≥96.0 %
- Specific optical rotation:-53° to -63° (20°C, 589 nm) (c=0.5, CHCl3)
Frequently Bought Together