Order Entry
Ireland
ContactUsLinkComponent
Trifluoromethanesulphonic anhydride for synthesis, Sigma-Aldrich®
Trifluoromethanesulphonic anhydride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Trifluoromethanesulphonic anhydride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    282.12 g/mol
  • Boiling Point:
    82 - 83 °C
  • Refractive Index:
    1.3210 - 1.3212 (20 °C, 589 nm)
  • Density:
    1.71 g/cm3 (20 °C)
  • Hill Formula:
    C2F6O5S2
  • Chemical Formula:
    (CF3SO2)2O

Specification Test Results

Related Information

Frequently Bought Together