

2,2,3,3,3-Pentafluoro-1-propanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:422-05-9
:
Formula:C₃H₃F₅O
MDL Number:MFCD00004673
:150.05 g/mol
Boiling Pt:78…80 °C (1013 hPa)
Density:1,5 g/cm³ (20 °C)
EINECS:207-012-7
UN:1986
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together