Order Entry
Ireland
ContactUsLinkComponent
Diammonium hydrogen citrate 99.0-103.0%, AnalaR NORMAPUR® ACS, Ph. Eur. analytical reagent
Diammonium hydrogen citrate 99.0-103.0%, AnalaR NORMAPUR® ACS, Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Diammonium hydrogen citrate 99.0-103.0%, AnalaR NORMAPUR® ACS, Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.0 - 103.0 %
  • C2O4 (Oxalate):
    Passes test
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Total S (as SO4):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • PO4 (Phosphate):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm

Specification Test Results

Frequently Bought Together