Diammonium hydrogen citrate ≥97%, GPR RECTAPUR®
Catalog # 21245.361
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bucket
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- Assay:Min. 97 %
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 0.1 %
- Fe (Iron):Max. 20 ppm
Specification Test Results
About this item
:
:226.19 g/mol
Merck Index:13,00513
Density:1,483 g/cm³ (20 °C)
EINECS:221-146-3
:GHS07
Storage Temperature:Ambient
CAS Number:3012-65-5
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068