Order Entry
Ireland
ContactUsLinkComponent
Diammonium hydrogen citrate
Diammonium hydrogen citrate
Catalog # MOLEM77018759
Supplier:  Molekula
CAS Number:  
Diammonium hydrogen citrate
Catalog # MOLEM77018759
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :226.19 g/mol
Merck Index:13,00513
Density:1,483 g/cm³ (20 °C)
EINECS:221-146-3
 :GHS07
Storage Temperature:Ambient
CAS Number:3012-65-5
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068