Diammonium hydrogen citrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3012-65-5
:
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068
:226.19 g/mol
Merck Index:14,512
Density:1,48
EINECS:221-146-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together