
Barium oxalate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:516-02-9
:
Formula:C₂BaO₄
MDL Number:MFCD00054429
:225.35 g/mol
Merck Index:14,985
Melting Pt:400 °C (dec.)
Density:2,658
Storage Temperature:Ambient
EINECS:208-216-9
UN:1564
ADR:6.1,II
:GHS07
Frequently Bought Together