Disodium oxalate
MOLE37145065-1KG
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
:
:134 g/mol
:13,08723
:250…270 °C
:2,27 g/cm³ (20 °C)
:200-550-3
:GHS07
:Ambient
:62-76-0
:Na₂C₂O₄
:MFCD00012465