Disodium oxalate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
CAS Number:62-76-0
:
Formula:Na₂C₂O₄
MDL Number:MFCD00012465
:134 g/mol
Merck Index:13,08723
Melting Pt:250…270 °C
Density:2,27 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:200-550-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:37 g/l (20 °C)
- Melting Point:250 - 270 °C (decomposition)
- Molar Mass:134 g/mol
- Bulk Density:600 kg/m3
- Density:2.27 g/cm3 (20 °C)
- pH:8 (30 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_106557
SDS
ProductPage_106557 ProductPage_106557
Product specification
Frequently Bought Together