Order Entry
Ireland
ContactUsLinkComponent
Succinic acid ≥99%, powder ACS
Irritant
Succinic acid ≥99%, powder ACS
Supplier:  MP Biomedicals
Certificates
undefined
Irritant
Succinic acid ≥99%, powder ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Assay:
    ≥99%
  • Melting range:
    185.0-191.0°C (w/i 2°C)
  • Insoluble matter:
    <0,01%
  • Residue on Ignition:
    <0,02%
  • Chloride :
    ≤0.001%
  • Phosphate:
    ≤0.001%
  • Sulphate:
    ≤0.003%
  • Nitrogen compounds:
    ≤0.001%
  • Heavy Metals:
    ≤5 ppm
  • Iron:
    ≤5 ppm
  • FTIR:
    Conforms to standard

Specification Test Results

Frequently Bought Together