

Mercaptoacetic acid (thioglycolic acid) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:68-11-1
:
Formula:HSCH₂COOH
MDL Number:MFCD00004876
:92.12 g/mol
Merck Index:13,09410
Boiling Pt:96 °C (1013 hPa)
Melting Pt:–16,5 °C
Density:1,33 g/cm³ (20 °C)
EINECS:200-677-4
UN:1940
ADR:8,II
:M-Clarity™ quality level = MQ200
Flash Pt:130 °C (closed cup)
:GHS06
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-16.5 °C
- Molar Mass:92.11 g/mol
- Boiling Point:220 °C (1013 hPa) (decomposition)
- Vapor Pressure:0.1 hPa (20 °C)
- Flash Point:131.5 °C
- Density:1.325 g/cm3 (20 °C)
- pH:1 (H2O, 20 °C)
Specification Test Results
Related Information
Frequently Bought Together