
Ammonium formate ≥99% for HPLC, Fluka™
Catalog # NOVS17843-50G
Supplier: Novartis
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:63.06 g/mol
Merck Index:13,00525
Melting Pt:119…121 °C
Density:1.28 g/cm³ (25 °C)
EINECS:208-753-9
:GHS07
Storage Temperature:Ambient
CAS Number:540-69-2
Formula:HCO₂NH₄
MDL Number:MFCD00013103
Density:1,28 g/cm³ (25 °C)