
Ammonium formate, (max. 3% H₂O) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-69-2
:
Formula:HCO₂NH₄
MDL Number:MFCD00013103
:63.06 g/mol
Merck Index:14,523
Melting Pt:∼116 °C
Density:1,28
EINECS:208-753-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together