Calcium stearate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Stearic acid calcium salt
CAS Number:1592-23-0
:66071-81-6
Formula:C₃₆H₇₀CaO₄
MDL Number:MFCD00036390
Density:1.065 g/cm³ (20 °C)
Molecular Weight:607.03 g/mol
Melting Pt:179…180 °C
Merck Index:13,01708
Storage Temperature:Ambient
EINECS:216-472-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together