

Sebacoyl dichloride for synthesis, Sigma-Aldrich®
: Merck
:111-19-3
:
:C₁₀H₁₆Cl₂O₂
:MFCD00000770
:239.14 g/mol
:220 °C (1013 hPa)
:–5 °C
:1,12 g/cm³ (20 °C)
:165 °C
:3265
:8,II
:M-Clarity™ quality level = MQ200
:203-843-4
:GHS05
:GHS07
:Ambient
- Solubility:(20 °C) (decomposition)
- Melting Point:-5 °C
- Molar Mass:239.14 g/mol
- Boiling Point:161 °C (19 hPa)
- Flash Point:113 °C
- Refractive Index:1.4684 (18 °C, 589 nm)
- Density:1.12 g/cm3 (20 °C)
- Synonyms:Sebacoyl chloride
- Hill Formula:C10H16Cl2O2
- Chemical Formula:ClOC(CH2)8COCl
Frequently Bought Together