Dimethyl oxalate
Catalog # MOLE41260542-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Oxalic acid dimethyl ester
:
:118.09 g/mol
Boiling Pt:163
Flash Pt:62
EINECS:209-053-6
UN:3261
:GHS07
Storage Temperature:Ambient
CAS Number:553-90-2
Formula:C₄H₆O₄
MDL Number:MFCD00008442
Melting Pt:54
Density:1.422 g/cm³ (15 °C)
Merck Index:12,06181