Dimethyl malonate ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-59-8
:
Formula:C₅H₈O₄
MDL Number:MFCD00008460
:132.12 g/mol
Merck Index:12,06169
Boiling Pt:181 °C (1013 hPa)
Melting Pt:–62 °C
Density:1,15 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-597-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (GC, area%):≥ 99.0 % (a/a)
- Density (d 20 °C/ 4 °C):1.151 - 1.155
- Identity (IR):passes test
- Boiling Point:181 - 183 °C (1013 hPa)
- Vapor Pressure:0.15 hPa (20 °C)
- Flash Point:85 °C
- Refractive Index:1.4135 (20 °C, 589 nm)
- Explosion Limit:1.3 - 17.4 %(V)
- Density:1.15 g/cm3 (20 °C)
- Ignition Point:440 °C
Specification Test Results
Frequently Bought Together