Bis(2-ethylhexyl)sebacate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-62-3
:
Formula:C₂₆H₅₀O₄
MDL Number:MFCD00009497
:426.68 g/mol
Merck Index:14,1251
Boiling Pt:211…213 °C (1 mmHg)
Melting Pt:–55 °C
Density:0,914
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:204-558-8
Related Information
Frequently Bought Together