Methyl stearate
Catalog # ROTH4704.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:298.51 g/mol
Merck Index:12,08959
Boiling Pt:355 °C (1013 hPa)
Melting Pt:37…39 °C
Density:1,007 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-990-4
CAS Number:112-61-8
Formula:CH₃(CH₂)₁₆CO₂CH₃
MDL Number:MFCD00009005