2'-Nitroacetanilide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:552-32-9
:
Formula:C₈H₈N₂O₃
MDL Number:MFCD00016991
:180.16 g/mol
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:209-009-6
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:90 - 93 °C
- Molar Mass:180.16 g/mol
- Hill Formula:C8H8N2O3
Specification Test Results
Frequently Bought Together