Order Entry
Ireland
Orders LinkContactUsLinkComponent
Hydroxyurea

Hydroxyurea

Glass bottle for solids, 25 g
Catalog # MOLEM18908038
Marketsource
Supplier:   Molekula
Hydroxyurea
Catalog # MOLEM18908038
Supplier:  Molekula
Marketsource
CAS Number:  127-07-1
Hydroxyurea

Glass bottle for solids, 25 g

CAS Number:  127-07-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:13,04874
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…144 °C
  • Storage Temperature:Ambient
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications