Hydroxyurea ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 127-07-1 Molecular Formula: CH4N2O2 Formula Weight: 76.06 Melting Point: ca 140° dec. Beilstein Reference: 1741548 Merck Reference: 14,4848 MDL No.: MFCD00007943
An inhibitor of DNA synthesis and an anticancer agent
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Merck Index:14,4848
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:∼ 140 °C (dec.)
- Storage Temperature:Ambient
- EINECS:204-821-7
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 127-07-1 Molecular Formula: CH4N2O2 Formula Weight: 76.06 Melting Point: ca 140° dec. Beilstein Reference: 1741548 Merck Reference: 14,4848 MDL No.: MFCD00007943
An inhibitor of DNA synthesis and an anticancer agent
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Merck Index:14,4848
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:∼ 140 °C (dec.)
- Storage Temperature:Ambient
- EINECS:204-821-7



