Order Entry
Ireland
ContactUsLinkComponent
Propionamide
Propionamide
Catalog # MOLE66160537-25G
Supplier:  Molekula
CAS Number:  
Propionamide
Catalog # MOLE66160537-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :73.09 g/mol
Merck Index:12,08009
Boiling Pt:213 °C (1013 hPa)
Melting Pt:79 °C
Density:1,1 g/cm³ (20 °C)
EINECS:201-172-1
CAS Number:79-05-0
Formula:C₃H₇NO
MDL Number:MFCD00008039