N,N-Dimethylformamide
Supplier: Roth Carl
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0,949 g/cm³ (20 °C)
Flash Pt:57,5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
Frequently Bought Together