
Diethyl pyrocarbonate, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:1609-47-8
:
Formula:O(COOC₂H₅)₂
MDL Number:MFCD00009106
:162.14 g/mol
Merck Index:13,08088
Boiling Pt:93 °C (1013 hPa)
Density:1,123 g/cm³ (20 °C)
Flash Pt:69 °C
EINECS:216-542-8
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together