
1-Chloro-3-methoxypropane for synthesis, Sigma-Aldrich®
8.43833.0025
: Merck
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Solubility:8 g/l (25 °C) (calculated)
- Molar Mass:108.57 g/mol
- Boiling Point:111 °C
- Vapor Pressure:25 hPa (20 °C)
- Flash Point:18 °C
- Density:0.999 g/cm3 (20 °C)
- Ignition Point:205 °C DIN 51794
- Hill Formula:C4H9ClO
:
:0,999 g/cm³ (20 °C)
:Ambient
:252-919-3
:1993
:3,II
:M-Clarity™ quality level = MQ200
:108.57 g/mol
:GHS02
:36215-07-3
:C₄H₉ClO
:MFCD05663419