Chloroform stabilised, ROTISOLV® Pestilyse®
ROTHT901.1
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
:
:119.38 g/mol
:13,02160
:61,7 °C (1013 hPa)
:–63 °C
:1,4832 g/cm³ (20 °C)
:Ambient
:200-663-8
:1888
:6.1,III
:Stabilised with ethanol
:05-2114614906-45
:GHS06
:GHS08
:67-66-3
:CHCl₃
:MFCD00000826