
10-Bromo-1-decanol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:53463-68-6
:
Formula:HO(CH₂)₉CH₂Br
MDL Number:MFCD00041681
:237.18 g/mol
Boiling Pt:165…169 °C (10 mmHg)
Density:1,190
Flash Pt:>110 °C (230 °F)
EINECS:258-572-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together