Order Entry
Ireland
ContactUsLinkComponent
1,10-Phenanthroline for synthesis, Sigma-Aldrich®
1,10-Phenanthroline for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
1,10-Phenanthroline for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    2.69 g/l (25 °C)
  • Melting Point:
    117 - 119 °C
  • Molar Mass:
    180.21 g/mol
  • Boiling Point:
    330 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together