

(S)-(-)-α-Methylbenzylamine for racemat separation, for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2627-86-3
:
Formula:C₈H₁₁N
MDL Number:MFCD00064406
:121.18 g/mol
Merck Index:12,06106
Boiling Pt:187 °C (1013 hPa)
Melting Pt:–10 °C
Density:0,95 g/cm³ (20 °C)
UN:2922
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
EINECS:220-098-0
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:42 g/l (20 °C)
- Melting Point:-10 °C
- Molar Mass:121.18 g/mol
- Boiling Point:187 °C (1013 hPa)
- Vapor Pressure:0.4 hPa (20 °C)
- Flash Point:71 °C
- Refractive Index:1.5259 (20 °C, 589 nm)
- Density:0.95 g/cm3 (20 °C)
- Ignition Point:355 °C
Specification Test Results
Frequently Bought Together